COM448677664 Combi-Blocks, Inc.

2-Chloro-6-phenoxypyrazine

  • MDL number MFCD00464983
  • Cas Number : 64383-29-5
  • Molecular Weight : 206.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8860

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8860
Availability details for
Quantity
  • Availability
  • Add to Cart