COM448677692 Combi-Blocks, Inc.

2-Cyano-3-(2-fluorophenyl)propionic acid

  • MDL number MFCD09800431
  • Cas Number : 948015-67-6
  • Molecular Weight : 193.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8903

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8903
Availability details for
Quantity
  • Availability
  • Add to Cart