COM448677718 Combi-Blocks, Inc.

2-Cyclopropylbenzonitrile

  • MDL number MFCD00463731
  • Cas Number : 3154-99-2
  • Molecular Weight : 143.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8938

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8938
Availability details for
Quantity
  • Availability
  • Add to Cart