COM448677724 Combi-Blocks, Inc.

2-Ethenyl-4-methoxy-pyridine

  • MDL number MFCD18384441
  • Cas Number : 30566-80-4
  • Molecular Weight : 135.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8946

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8946
Availability details for
Quantity
  • Availability
  • Add to Cart