COM448677740 Combi-Blocks, Inc.

2-Formyl-3-(4-fluorophenyl)-1-n-methyl indole

  • MDL number MFCD03839849
  • Cas Number : 93957-42-7
  • Molecular Weight : 253.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8973

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8973
Availability details for
Quantity
  • Availability
  • Add to Cart