COM448677743 Combi-Blocks, Inc.

2-Hexanamido-5-hydroxybenzoic acid

  • MDL number MFCD09998271
  • Cas Number : 1015856-35-5
  • Molecular Weight : 251.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8980

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8980
Availability details for
Quantity
  • Availability
  • Add to Cart