COM448677754 Combi-Blocks, Inc.

2-Hydroxy-2-(2-methoxyphenyl)ethylamine oxalate

  • MDL number MFCD04117771
  • Cas Number : 1171149-12-4
  • Molecular Weight : 257.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8996

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8996
Availability details for
Quantity
  • Availability
  • Add to Cart