COM448677774 Combi-Blocks, Inc.

2-Hydroxy-4-isopropyl-3-quinoline carboxylic acid methyl ester

  • MDL number MFCD00798202
  • Cas Number : 672310-22-4
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9017

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9017
Availability details for
Quantity
  • Availability
  • Add to Cart