COM448677812 Combi-Blocks, Inc.

2-Methoxy-4-(2-nitroethyl)phenol

  • MDL number MFCD03410699
  • Cas Number : 528594-30-1
  • Molecular Weight : 197.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9062

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9062
Availability details for
Quantity
  • Availability
  • Add to Cart