COM448677824 Combi-Blocks, Inc.

2-Methoxy-5-piperazin-1-yl-phenol

  • MDL number MFCD09261411
  • Cas Number : 180698-25-3
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9078

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9078
Availability details for
Quantity
  • Availability
  • Add to Cart