COM448677826 Combi-Blocks, Inc.

2-Methoxy-n-(prop-2-ynyl)acetamide

  • MDL number MFCD11100899
  • Cas Number : 537705-07-0
  • Molecular Weight : 127.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9081

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9081
Availability details for
Quantity
  • Availability
  • Add to Cart