COM448677835 Combi-Blocks, Inc.

2-Methyl-3-n,n-dimethylaminomethylindole

  • MDL number MFCD00022712
  • Cas Number : 37125-92-1
  • Molecular Weight : 188.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9099

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9099
Availability details for
Quantity
  • Availability
  • Add to Cart