COM448677869 Combi-Blocks, Inc.

2-Morpholinoisonicotinamide

  • MDL number MFCD11501090
  • Cas Number : 1086397-58-1
  • Molecular Weight : 207.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9148

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9148
Availability details for
Quantity
  • Availability
  • Add to Cart