COM448677871 Combi-Blocks, Inc.

2-N-(3-Aminopropyl)-amino-5-bromopyridine

  • MDL number MFCD07784021
  • Cas Number : 92993-40-3
  • Molecular Weight : 230.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9150

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9150
Availability details for
Quantity
  • Availability
  • Add to Cart