COM448677875 Combi-Blocks, Inc.

2-N-Boc-2-aminomethyl-3-(3-bromo-phenyl)-propionic acid

  • MDL number MFCD08061175
  • Cas Number : 886364-87-0
  • Molecular Weight : 358.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9155

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9155
Availability details for
Quantity
  • Availability
  • Add to Cart