COM448677883 Combi-Blocks, Inc.

2-N-Boc-amino-1-methylindole

  • MDL number MFCD11707036
  • Cas Number : 1159826-71-7
  • Molecular Weight : 246.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9165

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9165
Availability details for
Quantity
  • Availability
  • Add to Cart