COM448677886 Combi-Blocks, Inc.

2-N-Boc-amino-2-cyclopentyl-ethylamine

  • MDL number MFCD09261289
  • Cas Number : 936497-76-6
  • Molecular Weight : 228.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9168

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9168
Availability details for
Quantity
  • Availability
  • Add to Cart