COM448677889 Combi-Blocks, Inc.

2-N-Fmoc-aminomethyl piperidine

  • MDL number MFCD03001744
  • Cas Number : 672310-15-5
  • Molecular Weight : 336.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9172

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9172
Availability details for
Quantity
  • Availability
  • Add to Cart