COM448677915 Combi-Blocks, Inc.

2-Phenyl-3-pyrrolidin-1-yl-propan-1-ol

  • MDL number MFCD08061234
  • Cas Number : 889942-49-8
  • Molecular Weight : 205.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9214

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9214
Availability details for
Quantity
  • Availability
  • Add to Cart