COM448677936 Combi-Blocks, Inc.

2-Piperidin-4-yl-oxazole-4-carboxylic acid ethyl ester

  • MDL number MFCD02179430
  • Cas Number : 672310-05-3
  • Molecular Weight : 224.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9247

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9247
Availability details for
Quantity
  • Availability
  • Add to Cart