COM448677967 Combi-Blocks, Inc.

2-tert-Butoxycarbonylamino-4,4,4-trifluoro-3-methyl-butyric acid

  • MDL number MFCD02682485
  • Cas Number : 409333-54-6
  • Molecular Weight : 271.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9286

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9286
Availability details for
Quantity
  • Availability
  • Add to Cart