COM448678002 Combi-Blocks, Inc.

3-(1H-Imidazol-2-yl)-quinoline

  • MDL number MFCD08668961
  • Cas Number : 1006589-08-7
  • Molecular Weight : 195.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9332

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9332
Availability details for
Quantity
  • Availability
  • Add to Cart