COM448678005 Combi-Blocks, Inc.

3-(1-Trityl-1h-imidazol-4-yl)propan-1-amine

  • MDL number MFCD07784152
  • Cas Number : 195053-89-5
  • Molecular Weight : 367.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9336

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9336
Availability details for
Quantity
  • Availability
  • Add to Cart