COM448678007 Combi-Blocks, Inc.

3-(2-Amino-ethyl)-2-hydroxy-benzoic acid hydrobromide

  • MDL number MFCD17926324
  • Cas Number : 1255099-23-0
  • Molecular Weight : 262.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9340

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9340
Availability details for
Quantity
  • Availability
  • Add to Cart