COM448678009 Combi-Blocks, Inc.

3-(2-Aminoethyl)-5-chloroindolin-2-one, HCl

  • MDL number MFCD11100914
  • Cas Number : 1185150-35-9
  • Molecular Weight : 247.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9343

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9343
Availability details for
Quantity
  • Availability
  • Add to Cart