COM448678024 Combi-Blocks, Inc.

3-(2-Chlorophenyl)pyrrolidine oxalate

  • MDL number MFCD04117737
  • Cas Number : 1217062-15-1
  • Molecular Weight : 271.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9362

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9362
Availability details for
Quantity
  • Availability
  • Add to Cart