COM448678025 Combi-Blocks, Inc.

3-(2-Chlorophenyl)thiomorpholine

  • MDL number MFCD03002063
  • Cas Number : 887344-30-1
  • Molecular Weight : 213.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9363

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9363
Availability details for
Quantity
  • Availability
  • Add to Cart