COM448678026 Combi-Blocks, Inc.

3-(2-Chlorophenylthio)benzoic acid

  • MDL number MFCD09998244
  • Cas Number : 141358-05-6
  • Molecular Weight : 264.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9364

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9364
Availability details for
Quantity
  • Availability
  • Add to Cart