COM448678027 Combi-Blocks, Inc.

3-(2-Fluoro-phenyl)-piperazine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD08685980
  • Cas Number : 886767-09-5
  • Molecular Weight : 280.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9367

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9367
Availability details for
Quantity
  • Availability
  • Add to Cart