COM448678032 Combi-Blocks, Inc.

3(2H)-Isoquinolinone 7-bromo-1,4-dihydro-2-methyl-

  • MDL number MFCD16875464
  • Cas Number : 877265-10-6
  • Molecular Weight : 240.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9372

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9372
Availability details for
Quantity
  • Availability
  • Add to Cart