COM448678036 Combi-Blocks, Inc.

3-(2-Methoxybenzyl)pyrrolidine oxalate

  • MDL number MFCD04117742
  • Cas Number : 1170381-63-1
  • Molecular Weight : 281.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9377

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9377
Availability details for
Quantity
  • Availability
  • Add to Cart