COM448678057 Combi-Blocks, Inc.

3-(3,5-Dichlorophenyl)benzenesulfonyl chloride

  • MDL number MFCD01631883
  • Cas Number : 885950-92-5
  • Molecular Weight : 321.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9404

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9404
Availability details for
Quantity
  • Availability
  • Add to Cart