COM448678070 Combi-Blocks, Inc.

3-(3-Cyclopropyl-1-oxopropyl)-4(r)-(1-phenylmethyl)-2-oxazolidinone

  • MDL number MFCD02179407
  • Cas Number : 289677-11-8
  • Molecular Weight : 273.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9418

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9418
Availability details for
Quantity
  • Availability
  • Add to Cart