COM448678094 Combi-Blocks, Inc.

3-(4-Boc-amino-piperidin-1-yl)-2-methyl-propionic acid methyl ester

  • MDL number MFCD07369874
  • Cas Number : 886364-51-8
  • Molecular Weight : 300.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9458

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9458
Availability details for
Quantity
  • Availability
  • Add to Cart