COM448678100 Combi-Blocks, Inc.

3-(4-Bromophenyl)-dl-beta-alaninol hcl

  • MDL number MFCD06211739
  • Cas Number : 787615-14-9
  • Molecular Weight : 266.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9469

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9469
Availability details for
Quantity
  • Availability
  • Add to Cart