COM448678113 Combi-Blocks, Inc.

3-(4-Chlorobenzyl)pyrrolidine oxalate

  • MDL number MFCD04117740
  • Cas Number : 957998-82-2
  • Molecular Weight : 285.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9484

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9484
Availability details for
Quantity
  • Availability
  • Add to Cart