COM448678121 Combi-Blocks, Inc.

3-(4-Chlorophenyl)-dl-beta-alaninol, HCl

  • MDL number MFCD06211741
  • Cas Number : 1159824-48-2
  • Molecular Weight : 222.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9496

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9496
Availability details for
Quantity
  • Availability
  • Add to Cart