COM448678127 Combi-Blocks, Inc.

3-(4'-Fluorobenzoyl)indole

  • MDL number MFCD03839843
  • Cas Number : 152807-26-6
  • Molecular Weight : 239.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9504

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9504
Availability details for
Quantity
  • Availability
  • Add to Cart