COM448678129 Combi-Blocks, Inc.

3-(4-Fluorobenzyl)piperidine-3-ethylcarboxylate, HCl

  • MDL number MFCD03840059
  • Cas Number : 1189694-37-8
  • Molecular Weight : 301.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9506

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9506
Availability details for
Quantity
  • Availability
  • Add to Cart