COM448678155 Combi-Blocks, Inc.

3-(4-Fluorophenyl)thiomorpholine

  • MDL number MFCD03002061
  • Cas Number : 887344-28-7
  • Molecular Weight : 197.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9535

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9535
Availability details for
Quantity
  • Availability
  • Add to Cart