COM448678164 Combi-Blocks, Inc.

3-(4-Methoxybenzyl)pyrrolidine oxalate

  • MDL number MFCD04117739
  • Cas Number : 1188263-83-3
  • Molecular Weight : 281.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9544

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9544
Availability details for
Quantity
  • Availability
  • Add to Cart