COM448678192 Combi-Blocks, Inc.

3-(4-Methylbenzyl)piperidine-3-ethylcarboxylate oxalate

  • MDL number MFCD04117731
  • Cas Number : 1170556-67-8
  • Molecular Weight : 351.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9575

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9575
Availability details for
Quantity
  • Availability
  • Add to Cart