COM448678198 Combi-Blocks, Inc.

3-(4-Methylphenyl)benzenesulfonyl chloride

  • MDL number MFCD01631886
  • Cas Number : 885950-93-6
  • Molecular Weight : 266.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9581

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9581
Availability details for
Quantity
  • Availability
  • Add to Cart