COM448678207 Combi-Blocks, Inc.

3-(6-Chloro-3-pyridazinyloxy)phenylamine

  • MDL number MFCD11226193
  • Cas Number : 30184-98-6
  • Molecular Weight : 221.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9596

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9596
Availability details for
Quantity
  • Availability
  • Add to Cart