COM448678211 Combi-Blocks, Inc.

3-(Boc-amidino)-benzoic acid

  • MDL number MFCD05864634
  • Cas Number : 886363-46-8
  • Molecular Weight : 264.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9607

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9607
Availability details for
Quantity
  • Availability
  • Add to Cart