COM448678212 Combi-Blocks, Inc.

3-(Boc-amino)-4,5-dimethyl-1h-pyrazole

  • MDL number MFCD19382364
  • Cas Number : 1311254-41-7
  • Molecular Weight : 211.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9614

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9614
Availability details for
Quantity
  • Availability
  • Add to Cart