COM448678233 Combi-Blocks, Inc.

3-(Methylsulfonyl)pyrrolidine

  • MDL number MFCD01862542
  • Cas Number : 433980-62-2
  • Molecular Weight : 149.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9648

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9648
Availability details for
Quantity
  • Availability
  • Add to Cart