COM448678238 Combi-Blocks, Inc.

3-(Phenylsulfonyl)-6-oxa-3-azabicyclo[3.1.0]hexane

  • MDL number MFCD11040505
  • Cas Number : 955158-69-7
  • Molecular Weight : 225.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9655

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9655
Availability details for
Quantity
  • Availability
  • Add to Cart