COM448678262 Combi-Blocks, Inc.

3-(tert-Butylcarbamoyl)benzene-1-sulfonyl chloride

  • MDL number MFCD09805205
  • Cas Number : 409109-00-8
  • Molecular Weight : 275.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9686

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9686
Availability details for
Quantity
  • Availability
  • Add to Cart