COM448678319 Combi-Blocks, Inc.

3,4-Dichloroisothiazole-5-carbonitrile

  • MDL number MFCD09743745
  • Cas Number : 18480-52-9
  • Molecular Weight : 179.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9764

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9764
Availability details for
Quantity
  • Availability
  • Add to Cart